Learn More
PROCESS SOLUTIONS AUTOMATION & CONTROL PROCESS CONNECTION 1/4IN MALE
Catalog No. NC3360196
Encompass
Process Connection: 1/4" Male NPTWetted material: Hastelloy C276Pressure Range: 0 to 500 PSIPressure Reference: GaugeOutput Signal: 0-10V (3-wire)Electrical Connection: M12x1Options: NoneApprovals/Certifications: CE
Catalog No. NC3360196 Supplier PROCESS SOLUTIONS AUTOMATION & CONTROL Supplier No. CS102C00500PG7A00000
Process Connection: 1/4" Male NPTWetted material: Hastelloy C276Pressure Range: 0 to 500 PSIPressure Reference: GaugeOutput Signal: 0-10V (3-wire)Electrical Connection: M12x1Options: NoneApprovals/Certifications: CE