Learn More
Hach Company Process Head Holder TU5
Supplier:  Hach Company LZY946
Encompass
Process Head Holder for TU5300sc/TU5400scMaintenance of multiple turbidimeters can be quick and easy with the TU5 series process head holder The holder can be placed over the top of the instrument allowing for easy access of the instrument vial for cleaning and replacement
Catalog No. 50-002-07413