Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Process NPD™ One-Step Germicidal Detergent

Catalog No. 14415157 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
1 gal.
30 gal.
55 gal.
Container Type:
Drum
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Container Type
14-415-157 1 gal. Plastic Bottle
19-415-157 55 gal. Drum
14-415-158 30 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-415-157 Supplier STERIS Supplier No. 639008
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Process NPD™ one-step germicidal detergent is a quaternary ammonium disinfectant, a non-sterile, double bagged concentrate with no perfumes or dyes.

  • Non-sterile version of Process NPD™ family of one-step, quaternary ammonium cleaner/disinfectants made specifically for use on hard non-porous surfaces
  • Made with same formulation that contains no perfumes or dyes that might interfere with critical research studies or manufacturing processes
  • Use-dilution rate of 1:256
  • Kill a broad spectrum of common bacteria without harming surface

How Process NPD™ One-Step Germicidal Detergent Works

  • Liquid concentrate specifically formulated for disinfection of hard surfaces
  • Use-dilution can be applied manually or sprayed

Why Process NPD™ One-Step Germicidal Detergent?

  • Efficacy
    • Quaternary ammonium active ingredients provide broad spectrum antimicrobial efficacy
  • Productivity
    • Formulation includes surfactants for surface cleaning while disinfecting
  • Flexibility
    • Ideal for use in contamination control rotation
  • Reliability
    • Complete lot traceability
  • Assurance
    • Each lot documented by certificate of analysis

Specifications

Product Type Germicidal Detergent
pH 10.2 to 10.7
Sterility Non-sterile
Composition Quaternary ammonium
Container Type Plastic Bottle
Quantity 4 Pk.
Volume (English) 1 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.