Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Process NPD™ One-Step Germicidal Detergent

Catalog No. 19415157 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
1 gal.
30 gal.
55 gal.
Container Type:
Drum
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Container Type
19-415-157 55 gal. Drum
14-415-158 30 gal. Drum
14-415-157 1 gal. Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-415-157 Supplier STERIS Supplier No. 639001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Process NPD™ one-step germicidal detergent is a quaternary ammonium disinfectant, a non-sterile, double bagged concentrate with no perfumes or dyes.

This STERIS™ Process NPD™ One-Step Germicidal Detergent is the non-sterile version of the Process NPD™ family of one-step, quaternary ammonium cleaner/disinfectants made specifically for use on hard non-porous surfaces. The Process NPD™ products are all made with the same formulation that contains no perfumes or dyes that might interfere with critical research studies or manufacturing processes. At a use-dilution rate of 1:256, the Process NPD™ products kill a broad spectrum of common bacteria without harming the surface.

How Process NPD™ One-Step Germicidal Detergent Works

STERIS™ Process NPD™ One-Step Germicidal Detergent is a liquid concentrate specifically formulated for the disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.

Why Process NPD™ One-Step Germicidal Detergent?

  • Efficacy
    • Quaternary ammonium active ingredients provide broad spectrum antimicrobial efficacy.
  • Productivity
    • Formulation includes surfactants for surface cleaning while disinfecting.
  • Flexibility
    • Ideal for use in a contamination control rotation.
  • Reliability
    • Complete lot traceability.
  • Assurance
    • Each lot documented by Certificate of Analysis

Specifications

Product Type Germicidal Detergent
Form Concentrated Liquid
pH 10.2 to 10.7
Sterility Non-sterile
Container Type Drum
For Use With (Application) Cleaning and disinfecting washable hard, non-porous surfaces.
Volume (English) 55 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.