Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Process NPD™ st Sterile One-Step Germicidal Detergent

Catalog No. 14415156 Shop All STERIS Corporation Products
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
14-415-156 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-415-156 Supplier STERIS Supplier No. 160528
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Process NPD™ one-step germicidal detergent is a quaternary ammonium disinfectant, a sterile, double bagged concentrate with no perfumes or dyes.

  • Sterile version of Process NPD™ family of one-step, quaternary ammonium cleaner/disinfectants made specifically for use on hard non-porous surfaces
  • Made with same formulation that does not contain perfumes or dyes that might comprise critical manufacturing systems
  • Use-dilution rate of 1:256
  • Kill a broad spectrum of common bacteria without harming surface

How Process NPD™ st Sterile One-Step Germicidal Detergent Works

  • Liquid concentrate specifically formulated for disinfection of hard surfaces
  • Use-dilution can be applied manually or sprayed

Why Process NPD™ st Sterile One-Step Germicidal Detergent?

  • Efficacy
    • Quaternary ammonium active ingredients provide broad spectrum antimicrobial efficacy
  • Productivity
    • Double bagged and gamma irradiated for easy introduction into aseptic areas
  • Flexibility
    • Formulation includes surfactants for surface cleaning while disinfecting
  • Ease of Use
    • Ideal for use in a contamination control rotation
  • Assurance
    • Complete lot traceability and each lot documented by certificate of analysis

Specifications

Product Type Disinfectant
pH 10.2 to 10.7
Sterility Sterile
Composition Quaternary ammonium
Container Type Plastic Bottles
Quantity 144/Pk.
Volume (English) 1 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.