Learn More
Bellco Glass, Inc. The Bellco Process Controlled Overhead Drive is a versatile direct-drive agitator for cell culture and most aseptic mixing applications. The Overhead Drives were developed to eliminate the problem of magnetic agitator decoupling often observed in large v
SDP
Supplier:  Bellco Glass, Inc. 777420230
Encompass_Preferred
The Bellco Process Controlled Overhead Drive is a versatile direct-drive agitator for cell culture and most aseptic mixing applications. The Overhead Drives were developed to eliminate the problem of magnetic agitator decoupling often observed in large v
Catalog No. 50-194-6212