Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Hach Digital Online Process ORP Sensor, Insertion-Mount (DRD2P5)

Catalog No. NC2331897 Shop All Hach Company Products
Encompass

The DRD2P5 is a digital online insertion ORP sensor designed for continuous oxidation-reduction potential monitoring in process applications. It provides differential ORP measurement over a wide range and is intended for insertion-mount installations where reliable, long-term process monitoring is required.

  • Body material: PEEK.
  • Electrode material: Platinum (process electrode); titanium (ground electrode).
  • Measurement range: -1500 to +1500 mV ORP.
  • Cable length: 10 m (33 ft) standard.
  • Mounting type: Insertion.
  • Compatible controllers: SC digital controllers.
  • Model variations: Non-hazardous environment variant available.

Catalog No. NC2331897 Supplier Hach Company Supplier No. DRD2P5
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The DRD2P5 is a digital online insertion ORP sensor designed for continuous oxidation-reduction potential monitoring in process applications. It provides differential ORP measurement over a wide range and is intended for insertion-mount installations where reliable, long-term process monitoring is required.

  • Body material: PEEK.
  • Electrode material: Platinum (process electrode); titanium (ground electrode).
  • Measurement range: -1500 to +1500 mV ORP.
  • Cable length: 10 m (33 ft) standard.
  • Mounting type: Insertion.
  • Compatible controllers: SC digital controllers.
  • Model variations: Non-hazardous environment variant available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.