Learn More
Hach Company Process pH Electrolyte, 0.5 L, for APA series
Catalog No. NC3036734 Shop All Hach Company Products
Encompass
Process pH Electrolyte is a 0.5 L electrolyte solution formulated for use with APA series online analyzers to maintain pH electrode performance in continuous process monitoring applications.
  • Supplied in a 0.5 L bottle.
  • Formulated for pH electrode electrolyte fill and stabilization.
  • Designed for use with APA series online analyzers.
  • Helps maintain reliable pH measurement in continuous monitoring systems.
  • Store in a cool, dry place; keep containers tightly closed and protect from moisture.
Catalog No. NC3036734 Supplier Hach Company Supplier No. 2429149
Process pH Electrolyte is a 0.5 L electrolyte solution formulated for use with APA series online analyzers to maintain pH electrode performance in continuous process monitoring applications.
  • Supplied in a 0.5 L bottle.
  • Formulated for pH electrode electrolyte fill and stabilization.
  • Designed for use with APA series online analyzers.
  • Helps maintain reliable pH measurement in continuous monitoring systems.
  • Store in a cool, dry place; keep containers tightly closed and protect from moisture.