Learn More
Hach Company Process pH GP Insrt.Mnt. Sens.
Supplier:  Hach Company PD2P1A15P2S1N
Encompass
Hach pHD Online Process pH Sensor - General Purpose Insertion Mount pH SensorGeneral Purpose Online Process pH Sensor - pHD Technology Glass pH Electrode PEEK Housing Insertion Mount 4 5m Cable
Catalog No. 50-002-07497