Learn More
Hach Company Process pH GP Insrt.Mnt. Sens.
Catalog No. 5000207496 Shop All Hach Company Products
Encompass
Hach pHD Online Process pH Sensor - General Purpose Insertion Mount pH SensorGeneral Purpose Online Process pH Sensor - pHD Technology Glass pH Electrode PEEK Housing Insertion Mount 4 5m Cable
Catalog No. 50-002-07496 Supplier Hach Company Supplier No. PD2P1
Hach pHD Online Process pH Sensor - General Purpose Insertion Mount pH SensorGeneral Purpose Online Process pH Sensor - pHD Technology Glass pH Electrode PEEK Housing Insertion Mount 4 5m Cable