Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ Process QDS Disinfectant Cleaner

Catalog No. 19999574 Shop All STERIS Corporation Products
Change view
Click to view available options
Size:
4 x 1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-999-574 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-999-574 Supplier STERIS™ Supplier No. 1J7508
Add to Cart
Add to Cart

Process QDS Process and Research Cleaner is a phosphate-free, one-step cleaner/disinfectant formulated for use on hard non-porous surfaces. This product is recommended for use in disinfecting, sanitizing, and deodorizing hard non-porous surfaces. It is effective in hard water and organic soil loads.

STERIS' Process QDS Process and Research Cleaner is a phosphate-free, one-step cleaner/disinfectant formulated for use on hard non-porous surfaces. This product is recommended for use in disinfecting, sanitizing, and deodorizing hard non-porous surfaces. It is effective in hard water and organic soil loads.

HOW PROCESS QDS PROCESS AND RESEARCH CLEANER WORKS

STERIS' Process QDS Process and Research Cleaner is a liquid concentrate specifically formulated for the disinfection of hard non-porous surfaces. The use-dilution can be applied manually or sprayed.

WHY PROCESS QDS PROCESS AND RESEARCH CLEANER?

  • Efficacy - Quaternary ammonium active ingredients provide broad spectrum antimicrobial efficacy.
  • Productivity - Formulation includes surfactants for surface cleaning while disinfecting.
  • Reliability - Ideal for use in a contamination control rotation.
  • Flexibility - Can be used as a disinfectant or sanitizer.
  • Assurance - Complete lot traceability with each lot documented by Certificate of Analysis.

Specifications

Product Type Cleaner/Disinfectant
Form Clear Liquid
pH 6.1-8.8 (Undiluted)
Scent Unscented
Sterility Non-sterile
Composition Quaternary ammonium
Size 4 x 1 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.