Learn More
Comecerbrand Process Qualification Valve for Flange PQ hole 100 μm
Catalog No. 17980821
Change view
Click to view available options
Product Type:
Valve
Catalog No. Product Type
17-980-821 Valve
1 options
Catalog No. 17-980-821 Supplier Comecerbrand Supplier No. NL0254020002
Add to Cart
Add to Cart
Process qualification valve used for testing and verifying SpeedyGlove performance through a calibrated opening. OPTION.
  • Process qualification valve with a calibrated 100 μm hole for PQ of the SpeedyGlove.
  • Serves as the standard valve for performing process qualification.
  • Required to complete PQ and generate an accurate testing recipe.
  • One PQ valve of any type, based on process needs, is necessary for proper qualification.
Specifications
For Use With (Equipment) Glove Integrity Tester
Product Type Valve