Learn More
Comecerbrand Process Qualification Valve for Flange PQ hole 250 μm
Catalog No. 17980824
Change view
Click to view available options
Product Type:
Valve
Catalog No. Product Type
17-980-824 Valve
1 options
Catalog No. 17-980-824 Supplier Comecerbrand Supplier No. NL0254020008
Add to Cart
Add to Cart
Process qualification valve used for testing and verifying SpeedyGlove performance through a calibrated opening. OPTION.
  • Process qualification valve with a calibrated 250 μm hole for PQ testing.
  • Used to support the PQ process for the SpeedyGlove.
  • Required to perform PQ and generate an accurate testing recipe.
  • One PQ valve of any type, based on process needs, is necessary for proper qualification.
Specifications
For Use With (Equipment) Glove Integrity Tester
Product Type Valve