Learn More
SMART TUBE INC Processing Inserts, Bag of 100
Catalog No. 501351698
Encompass_Preferred
A bag of one hundred (100) Processing Inserts. Processing Inserts are used after thawing samples previously frozen in Smart Tubes. The Processing Insert is used to decant a blood samples from a Smart Tube. User information must be added to all orders for accurate processing
This item is not returnable. View return policy
Catalog No. 501351698 Supplier SMART TUBE INC Supplier No. SERT1
A bag of one hundred (100) Processing Inserts. Processing Inserts are used after thawing samples previously frozen in Smart Tubes. The Processing Insert is used to decant a blood samples from a Smart Tube. User information must be added to all orders for accurate processing