Learn More
MESA LABS BOZEMAN MFG FACILITY BOWIE DICK TEST PACK 20/BX
SDP
Catalog No. NC1949523
Encompass
BOWIE DICK TYPE TESTS - PROCHEM BD TYPE 2. Preassembled, disposable test pack. Distinct, non-reversible pink to black color change. 3.5 minutes at 132°C in a pre-vacuum steam sterilizer. Conforms to applicable portions of ANSI/AMMI/ISO 11140-5. Box of 20 test packs. 3 year expiry.
Catalog No. NC1949523 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. 7705CIBDTLF20
BOWIE DICK TYPE TESTS - PROCHEM BD TYPE 2. Preassembled, disposable test pack. Distinct, non-reversible pink to black color change. 3.5 minutes at 132°C in a pre-vacuum steam sterilizer. Conforms to applicable portions of ANSI/AMMI/ISO 11140-5. Box of 20 test packs. 3 year expiry.