Learn More
MESA LABS BOZEMAN MFG FACILITY Mesa Labs ProChem BD Bowie-Dick Test Pack, Pre-Vacuum Steam Sterilizer Monitoring, 80 Test Packs/Cs
SDP
Catalog No. NC0534444
Encompass
The Mesa Labs ProChem BD Bowie-Dick Type Air Removal Test Pack offers a reliable solution for daily monitoring of pre-vacuum steam sterilizers. This disposable, pre-assembled test pack ensures effective steam penetration with its distinct pink to black color change, indicating air removal performance. Designed for safety with lead-free ink, it complies with international standards for quality in sterilization processes.
This item is not returnable. View return policy
Catalog No. NC0534444 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. 7705CIBDTLF
The Mesa Labs ProChem BD Bowie-Dick Type Air Removal Test Pack offers a reliable solution for daily monitoring of pre-vacuum steam sterilizers. This disposable, pre-assembled test pack ensures effective steam penetration with its distinct pink to black color change, indicating air removal performance. Designed for safety with lead-free ink, it complies with international standards for quality in sterilization processes.