Learn More
Medchemexpress LLC HY-B0845 250mg Medchemexpress, Prochloraz CAS:67747-09-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0845250MG
Encompass_Preferred
Medchemexpress, HY-B0845 250mg Prochloraz CAS:67747-09-5 Prochloraz is an imidazole antifungal that inhibits ergosterol biosynthesis via inhibition of the cytochrome P450-dependent 14α-demethylation of lanosterol, which results in disruption of the fungal cell membrane and cell death. Prochloraz inhibits human placenta microsomal aromatase in vitro (IC50=40 nM). Prochloraz also acts as an antagonist of the estrogen receptor (ER) and androgen receptor (AR) (IC50s=25 μM and 4 μM, respectively) as well as activates the aryl hydrocarbon receptor (AhR; EC50=1 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2663