Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prochloraz 98.0+%, TCI America™
SDP

Catalog No. P21371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21371G 1 g
P21375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21371G Supplier TCI America Supplier No. P21371G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67747-09-5
Molecular Formula C15H16Cl3N3O2
Molecular Weight (g/mol) 376.662
MDL Number MFCD00078735
InChI Key TVLSRXXIMLFWEO-UHFFFAOYSA-N
Synonym prochloraz, mirage, octave, prelude, ascurit, sporgon, sportak, mirage pesticide, dibavit, sprint
PubChem CID 73665
ChEBI CHEBI:8434
IUPAC Name N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]imidazole-1-carboxamide
SMILES CCCN(CCOC1=C(C=C(C=C1Cl)Cl)Cl)C(=O)N2C=CN=C2

Specifications

Melting Point 50°C
Quantity 1 g
UN Number 3077
Formula Weight 376.66
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Prochloraz
RTECSNumber NI4000400
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.