Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Procion blue HB | 12236-82-7 | MFCD00043102 | 97.9% | 774.16 g/mol | C29H20ClN7O11S3 | 10 MG
SDP

Catalog No. 5000299296
Encompass_Preferred

Procion Blue HB (Reactive Blue 2) is a triazine dye and purinergic antagonist used in pharmacological research and biochemical assays. It is supplied for research use with reported high purity and is offered in small laboratory pack sizes for assay development and signaling studies.

  • Purity 97.9%.
  • CAS number 12236-82-7.
  • Molecular weight 774.16 g/mol.
  • Soluble in DMSO, water, and ethanol (laboratory solvents).
  • Suitable for studies of ATP receptors and calcium signaling assays.

Catalog No. 50-002-99296 Supplier Medchemexpress LLC Supplier No. HYD096510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Procion Blue HB (Reactive Blue 2) is a triazine dye and purinergic antagonist used in pharmacological research and biochemical assays. It is supplied for research use with reported high purity and is offered in small laboratory pack sizes for assay development and signaling studies.

  • Purity 97.9%.
  • CAS number 12236-82-7.
  • Molecular weight 774.16 g/mol.
  • Soluble in DMSO, water, and ethanol (laboratory solvents).
  • Suitable for studies of ATP receptors and calcium signaling assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.