Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gaylord Chemical Company, LLC PROCIPIENT™ (Dimethyl Sulfoxide USP, PhEur)
GREENER_CHOICESDP

Catalog No. 18614921 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-614-921 4 L
18-614-920 6 x 1 L
18-614-917 4 x 4 L
18-614-918 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-614-921 Supplier Gaylord Chemical Company, LLC Supplier No. 5010450LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Procipient™ DMSO is both an active pharmaceutical ingredient and a versatile excipient that is widely used in various pharmaceutical and biotechnology applications. Procipient is an extremely polar, water-soluble solvent with a minimum purity of 99.99%.

Procipient™ Characteristics:

  • Highly purified DMSO with a minimum purity of 99.99%
  • Water miscible/extremely polar solvent
  • Odorless, clear water-white liquid
  • Essentially non-toxic by all routes of administration
  • Chemically compatible with other excipients
  • Low viscosity (2.0 cP at 25°C)
  • Density similar to water
  • (1.0955 g / mL at 25°C)
  • Low volatility: 189°C boiling point

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.5°C (65.3°F)
Color Clear
Density 1.0955 g/mL (25°C)
Quantity 4 L
Flash Point 87°C (188.6°F) (Closed Cup)
Ignition Point 215°C (419°F)
Odor Odorless
Physical Form Liquid
Vapor Pressure 59.4 Pa at 20°C (68°F)
Viscosity 2.0 cP (25°C)
Boiling Point 189°C
Solubility Information Water Miscible, Extremely Polar Solvent
Percent Purity 99.99%
Grade USP
Chemical Name or Material Dimethyl Sulfoxide
Show More Show Less
Recommended Storage Store in a well-ventilated place. Keep cool.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.