Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chembio Procise ADL Assay Adalimumab
SDP

Catalog No. 21010245
Change view
Click to view available options
Form:
Lyophilized
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-010-245 Lyophilized
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-010-245 Supplier Chembio Supplier No. 5430
Add to Cart
Add to Cart
This item is not returnable. View return policy

Procise ADL™ is a rapid, quantitative assay for therapeutic drug monitoring of adalimumab in serum. Used with the ProciseDx™ Analyzer, it supports IBD treatment decisions with results in under 5 minutes.

  • Quantitative TDM for Adalimumab: Enables rapid measurement of adalimumab levels in serum to support therapeutic management of IBD patients.
  • Fast Results: Delivers accurate, quantitative results in under 5 minutes using the ProciseDx™ Analyzer.
  • Validated for Clinical Use: FDA-cleared for use in moderate-complexity labs; supports both adult and pediatric Crohn's disease and ulcerative colitis care.
  • Proven Analytical Performance: Sensitive and specific TR-FRET-based immunoassay with a reportable range of 1.0 to 50 μg/mL.
  • Simple Workflow: Uses standard serum samples and minimal hands-on time.

Specifications

Certifications/Compliance FDA DeNovo
Content And Storage 15°C to 30°C (59°F to 86°F)
Control Sets Included High and Low
CE Marker Yes
Clia Complexity Moderate (Prescription Use Only)
Detectable Analytes Adalimumab
Detection Method Measures Relative Fluorescence Units (RFU)
DoA Calibrators No
Sample Type Serum
Type Test Cartridge
Disposable Single-use
Level High, Low
Analysis Time <5 minutes
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.