Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Procise™ cLC Flask Maintenance Kit

Catalog No. 401990
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-199-0 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-199-0 Supplier Applied Biosystems™ Supplier No. 401990
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This is a maintenance accessory for the Procise™ system.

  • For Research Use Only
  • Not for use in diagnostics procedures
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Procise Protein Sequencer
Product Type cLC Flask Maintenance Kit
Product Line Procise
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Format Kit
Quantity 1 kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.