Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chembio ProciseDx Calibration Cart
SDP

Catalog No. 21010244
Change view
Click to view available options
Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-010-244 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-010-244 Supplier Chembio Supplier No. 5429
Add to Cart
Add to Cart
This item is not returnable. View return policy

The ProciseDx™ Calibration Cartridge confirms the analyzer is functioning within specifications. It uses embedded fluorescence values and is required for system calibration every 30 days.

  • Used to verify the ProciseDx™ Analyzer is operating within its defined specifications
  • Contains embedded values that are read by the analyzer's optical system to confirm performance
  • Calibration is required every 30 days as indicated by the analyzer; the cartridge must be used to complete this calibration procedure

Specifications

Content And Storage 15°C to 30°C (59°F to 86°F)
For Use With (Equipment) ProciseDx Analyzer
Sample Type Serum
Type Test Cartridge
Disposable Reusable
Includes 1 Calibration Cartridge, Product Insert
Level Normal
Quantity 1 Ea.
Sterility Non-sterile
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.