Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProClean™ 1 Beard Cover

Catalog No. 19088874 Shop All DuPont Personal Protection Products
Change view
Click to view available options
ProClean™ 1 Beard Cover
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-088-874 ProClean™ 1 Beard Cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-088-874 Supplier DuPont™ Supplier No. GA793SWH0003000B

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

DuPont™ ProClean™ Apparel for Cleanrooms

ProClean™ garments are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other cleanroom or controlled critical environments.

TRUSTED_SUSTAINABILITY

Specifications

Type Cleanroom Beard Cover
Size Universal
Color White
Disposable Single-use
For Use With (Application) Cleanrooms
Packaging Quantity 300/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.