Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProClean™ 1 Bouffant

Catalog No. 19088875 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Color:
White
Size:
21 in.
24 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-088-875 White 21 in.
19-088-876 White 24 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-088-875 Supplier DuPont™ Supplier No. GA729SWH0003000B
This item is not returnable. View return policy

DuPont™ ProClean™ bouffants are used in cleanroom applications.

ProClean™ garments are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other cleanroom or controlled critical environments.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Bouffant
Size 21 in.
Color White
Disposable Single-use
Model GA729SWH0003000B
For Use With (Application) Controlled Environments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.