Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProClean™ 1 Bouffant

Catalog No. 19088876 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Color:
White
Size:
21 in.
24 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-088-876 White 24 in.
19-088-875 White 21 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-088-876 Supplier DuPont™ Supplier No. GA728SWH0003000B

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DuPont™ ProClean™ bouffants are used in cleanroom applications.

ProClean™ garments are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other cleanroom or controlled critical environments.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Bouffant
Size 24 in.
Color White
Disposable Single-use
Model GA728SWH0003000B
For Use With (Application) Controlled Environments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.