Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProClean™ 2 Sterilization Wrap, 54 x 54 in.

Catalog No. 19088833 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Dimensions (L x W):
54 x 54 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-088-833 54 x 54 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-088-833 Supplier DuPont™ Supplier No. GB0540BU0001000B
This item is not returnable. View return policy

DuPont™ ProClean™ Apparel for Cleanrooms

ProClean™ garments are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other cleanroom or controlled critical environments.

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
For Use With (Application) Controlled Environment
Dimensions (L x W) 54 x 54 in.
Product Type Sterilization Wrap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.