Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProClean™ Coverall (all)

Catalog No. 19088359 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Height (English):
5.11 to 6.3 ft.
5.5 to 5.10 ft.
5.8 to 6 ft.
6.1 to 6.5 ft.
6.2 to 6.6 ft.
6.3 to 7 ft.
Height (Metric):
155.75 to 192.04 cm
167.64 to 155.44 cm
176.78 to 182.88 cm
185.92 to 198.12 cm
188.97 to 201.16 cm
192.02 to 213.3 cm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English) Height (Metric)
19-088-359 3X-Large 6.2 to 6.6 ft. 188.97 to 201.16 cm
19-088-355 Medium 5.5 to 5.10 ft. 167.64 to 155.44 cm
19-088-358 2X-Large 6.1 to 6.5 ft. 185.92 to 198.12 cm
19-088-360 4X-Large 6.3 to 7 ft. 192.02 to 213.3 cm
19-088-357 X-Large 5.11 to 6.3 ft. 155.75 to 192.04 cm
19-088-356 Large 5.8 to 6 ft. 176.78 to 182.88 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-088-359 Supplier DuPont™ Supplier No. PC143SWH3X00250B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Unique breathable composite material provides particulate and light splash protection for critical environments. Coveralls feature a zipper front, serged seams, and elastic wrists, ankles, and waist.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Coverall has elastic openings for tighter fit at wrist and ankle.
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing.
  • Enclosed elastic at the back of the garment for a better fit
  • Bulk packaged in double transparent poly liners.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • D13477891,D13477875,D13477852,D13477915,D13477791,D13477836
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size 3X-Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Back Style Elastic
Size (English) Chest 46 to 52 in.
Size (English) Inseam 32.5 in.
Size (Metric) Chest 116.8 to 132.08 cm
Size (Metric) Inseam 82.55 cm
Sleeve Style Set
Thickness 5.4 mil
For Use With (Application) Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments
Length (English) Sleeve 40.5 in.
Length (Metric) Sleeve 102.87 cm
Height (English) 6.2 to 6.6 ft.
Height (Metric) 188.97 to 201.16 cm
Packaging Bulk
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.