Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProClean™ Limited-Use Cleanroom Coverall with Hood and Boots

Catalog No. 190881592 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-088-1592 2X-Large
19-088-1591 Large
19-088-1590 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-088-1592 Supplier DuPont™ Supplier No. PC122SWH2X00250C

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for cleanroom operations

  • Full-length zippered front
  • Attached hood and boots with elastic face opening
  • Tunnelized, snug-fitting elastic at the face, wrists, ankles, and back-waist

Order Info

Available with no special cleaning (bulk) or cleaned, in a variety of sizes.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size 2X-Large
Material Tyvek
Color White
Disposable Single-use
Sterility Non-sterile
Description Finish: Clean
For Use With (Application) Controlled Environments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.