Learn More
Supelco Inc ProClinA 150, pkg of 50A mL
Catalog No. NC9370406
Encompass
ProClin products from MilliporeSigma are only labelled for use in In Vitro Diagnostics (IVD): “in vitro diagnostics are tests that can detect diseases, conditions, or infections.”
Catalog No. NC9370406 Supplier Supelco Inc Supplier No. 49376U
ProClin products from MilliporeSigma are only labelled for use in In Vitro Diagnostics (IVD): “in vitro diagnostics are tests that can detect diseases, conditions, or infections.”