Learn More
Biomatik Corporation Procollagen Iii N-Terminal Propeptide (Piiinp) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU30200
Encompass_Preferred
Procollagen Iii N-Terminal Propeptide (Piiinp) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Procollagen III N-Terminal Propeptide (PIIINP). The Procollagen Iii N-Terminal Propeptide (Piiinp) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Human (Homo sapiens). Applications: WB. Target Synonyms: Procollagen III N-Terminal Propeptide (PIIINP). Accession Number: P02461. Gene ID: 1281. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4767