Learn More
REPLIGEN CORPORATION ProConnex® TFF Self Assembly Kit for KR2i - MICRO - 15/50ML BTL
SDP
Catalog No. NC3936486
Encompass
This ProConnex® TFF Self Assembly Kit is designed for use with the KrosFlo® KR2i TFF System and handles working volumes ranging from 0.75mL to 50mL, depending on the bottle used. Compatible with Spectrum® Hollow Fiber MicroKros and MidiKros Modules.
Catalog No. NC3936486 Supplier REPLIGEN CORPORATION Supplier No. SKITCD17490N
This ProConnex® TFF Self Assembly Kit is designed for use with the KrosFlo® KR2i TFF System and handles working volumes ranging from 0.75mL to 50mL, depending on the bottle used. Compatible with Spectrum® Hollow Fiber MicroKros and MidiKros Modules.