Learn More
ACROBiosystems Recombinant Protein;Human PROCR / EPCR / CD201 Protein, Fc Tag;HEK293;1MG;PRR,PROCR,EPCR,CD201,APC receptor,CCCA,CCD41
Catalog No. 501658176
Encompass_Preferred
Recombinant Protein;Human PROCR / EPCR / CD201 Protein, Fc Tag;1MG;Human PROCR, Fc Tag (PRR-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 18 - Thr 209 (Accession # AAH14451).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.6 kDa. As a result of glycosylation, the protein migrates as 65 kDa under reducing (R) condition, and 125 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-8176 Supplier ACROBiosystems Supplier No. PRRH52541MG
Recombinant Protein;Human PROCR / EPCR / CD201 Protein, Fc Tag;1MG;Human PROCR, Fc Tag (PRR-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 18 - Thr 209 (Accession # AAH14451).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.6 kDa. As a result of glycosylation, the protein migrates as 65 kDa under reducing (R) condition, and 125 kDa under non-reducing (NR) condition (SDS-PAGE).