Learn More
Synbiosis™ ProcScan
GREENER_CHOICESDP
Catalog No. 01257329 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Includes:
ProtoCOL 3 Software, Scanner
Scanner Only
For Use With (Equipment):
ProtoCOL 3 Instruments
ProtoCOL 3 Instruments or Standalone
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes For Use With (Equipment)
01-257-329 ProtoCOL 3 Software, Scanner ProtoCOL 3 Instruments or Standalone
01-257-330 Scanner Only ProtoCOL 3 Instruments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-257-329 Supplier Synbiosis™ Supplier No. PROCSCAN
Add to Cart
Add to Cart
Request a Quote
ProcScan is designed to accommodate plates too large to fit inside the ProtoCOL 3 for colony counting or for the measurement of inhibition zones.
  • For large size plate: Using the ProScan enables you to use all the features of the ProtoCOL 3 software but on larger size samples, working with plates up to 300 x 300 mm
  • Connect directly to a ProtoCOL 3 or to a stand alone PC with ProtoCOL 3 software
  • 12 month warranty
Specifications
Product Type Zone Imaging, Plate Imaging
Material Plastic-molded components
Outlet Style Standard USA Type B Plug
Product Line Synbiosis
Sample Size 300 x 300 mm
For Use With (Application) Automatic colony counting and zone measuring
For Use With (Equipment) ProtoCOL 3 Instruments or Standalone
Includes ProtoCOL 3 Software, Scanner