Description
- For large size plate: Using the ProScan enables you to use all the features of the ProtoCOL 3 software but on larger size samples, working with plates up to 300 x 300 mm
- Connect directly to a ProtoCOL 3 or to a stand alone PC with ProtoCOL 3 software
- 12 month warranty
Specifications
Specifications
| Product Type | Zone Imaging, Plate Imaging |
| Material | Plastic-molded components |
| Outlet Style | Standard USA Type B Plug |
| Product Line | Synbiosis |
| Sample Size | 300 x 300 mm |
| For Use With (Application) | Automatic colony counting and zone measuring |
| For Use With (Equipment) | ProtoCOL 3 Instruments |
| Includes | Scanner Only |