Learn More
Enterprise Technology Solutions Procter & Gamble Commercial Degreaser, Heavy-duty, Dawn, 1 Gal, 3/CT, Purple
SDP
Catalog No. 5000790348
Encompass_Preferred
Dawn Professional Heavy-Duty Degreaser cuts through tough jobs with the grease-cutting power of Dawn. It is designed to clean your toughest grease problems. Use full-strength to clean tough, greasy buildup on range hoods, fryers, ovens and walls. It is safe for use on food-contact surfaces when followed by rinsing with water. This heavy-duty degreaser with a pine scent is excellent for cleaning corral areas and dumpsters as well as waxed tile floors without damaging the wax when properly diluted and spot-cleaning where heavy grease buildups exist. Also use to clean heavy crank case oil from concrete drive-thru pads and sidewalks. Noncaustic, nonflammable formula contains no phosphates.
Catalog No. 50-007-90348 Supplier Enterprise Technology Solutions Supplier No. PGC04852
Dawn Professional Heavy-Duty Degreaser cuts through tough jobs with the grease-cutting power of Dawn. It is designed to clean your toughest grease problems. Use full-strength to clean tough, greasy buildup on range hoods, fryers, ovens and walls. It is safe for use on food-contact surfaces when followed by rinsing with water. This heavy-duty degreaser with a pine scent is excellent for cleaning corral areas and dumpsters as well as waxed tile floors without damaging the wax when properly diluted and spot-cleaning where heavy grease buildups exist. Also use to clean heavy crank case oil from concrete drive-thru pads and sidewalks. Noncaustic, nonflammable formula contains no phosphates.