Learn More
Enterprise Technology Solutions Procter & Gamble Commercial Degreaser,Hvy-Dty,Pro,Multi-Surface,RTU,Daw,32oz,Purple
SDP
Catalog No. 5000829845
Encompass_Preferred
Dawn Professional Multi-Surface Heavy Duty Degreaser Spray cleans your toughest grease problems. It can be used full-strength to clean tough, greasy buildup on range hoods, fryers, ovens and walls. Spray is safe for use on food-contact surfaces when followed by rinsing with water. It is excellent for cleaning corral areas and dumpsters. Use for cleaning heavy crank case oil from concrete drive-thru pads and sidewalks. Also clean waxed tile floors without damaging the wax when properly diluted and spot-cleaning where heavy grease buildups exist. Formulated for employee safety, degreaser spray contains no phosphate and is noncaustic and nonflammable.
Catalog No. 50-008-29845 Supplier Enterprise Technology Solutions Supplier No. PGC07308
Dawn Professional Multi-Surface Heavy Duty Degreaser Spray cleans your toughest grease problems. It can be used full-strength to clean tough, greasy buildup on range hoods, fryers, ovens and walls. Spray is safe for use on food-contact surfaces when followed by rinsing with water. It is excellent for cleaning corral areas and dumpsters. Use for cleaning heavy crank case oil from concrete drive-thru pads and sidewalks. Also clean waxed tile floors without damaging the wax when properly diluted and spot-cleaning where heavy grease buildups exist. Formulated for employee safety, degreaser spray contains no phosphate and is noncaustic and nonflammable.