Learn More
Enterprise Technology Solutions Procter & Gamble Commercial Dish Detergent,Powerwash,Heavy-Duty,Spray,16oz,BE
SDP
Catalog No. 5000790708
Encompass_Preferred
Dawn Professional Heavy Duty Powerwash is a must-have in every foodservice and restaurant kitchen. It's perfect for washing all wares outside of the three-compartment sink. Clean hard-to-wash, immobile or bulky items such as deli slicers, stationary mixers and other larger wares. Pre-treat tough, baked-on grease before washing in your three-compartment sink. Quickly clean specialty and frequently used items. With the power of an 8-hour soak in minutes, Dawn Professional Powerwash helps break down tough commercial grease fast. Waterless activation dissolves grease before you start scrubbing. Formula includes three cleaning technologies not found in traditional pot and pan detergents.
Catalog No. 50-007-90708 Supplier Enterprise Technology Solutions Supplier No. PGC12301
Dawn Professional Heavy Duty Powerwash is a must-have in every foodservice and restaurant kitchen. It's perfect for washing all wares outside of the three-compartment sink. Clean hard-to-wash, immobile or bulky items such as deli slicers, stationary mixers and other larger wares. Pre-treat tough, baked-on grease before washing in your three-compartment sink. Quickly clean specialty and frequently used items. With the power of an 8-hour soak in minutes, Dawn Professional Powerwash helps break down tough commercial grease fast. Waterless activation dissolves grease before you start scrubbing. Formula includes three cleaning technologies not found in traditional pot and pan detergents.