Learn More
Enterprise Technology Solutions Procter & Gamble Commercial Dishwasher Detergent,ActonPacs,Platinum,w/Dawn,FreshScent,59/CT,Green
SDP
Catalog No. 5000790519
Encompass_Preferred
Cascade Platinum ActionPacs dishwasher detergent scrubs away dried on food with no pre-washing required. The pod dissolves fast to start cleaning right away, releasing the soaking power of Dawn dishwashing liquid, while food-seeking enzymes latch on and break down food into particles so small they can flow right down the drain. Cascade Platinum dishwashing detergent is formulated to remove visible and invisible residue for gleaming glassware and dazzling dishes. Save up to 20 gallons of water per dishwasher load when you skip the pre-wash and run your dishwasher with Cascade Platinum.
Catalog No. 50-007-90519 Supplier Enterprise Technology Solutions Supplier No. PGC15534
Cascade Platinum ActionPacs dishwasher detergent scrubs away dried on food with no pre-washing required. The pod dissolves fast to start cleaning right away, releasing the soaking power of Dawn dishwashing liquid, while food-seeking enzymes latch on and break down food into particles so small they can flow right down the drain. Cascade Platinum dishwashing detergent is formulated to remove visible and invisible residue for gleaming glassware and dazzling dishes. Save up to 20 gallons of water per dishwasher load when you skip the pre-wash and run your dishwasher with Cascade Platinum.