Learn More
Enterprise Technology Solutions Procter & Gamble Commercial Dishwashing Liquid, Original, 1 Gallon, Blue
SDP
Catalog No. 5000790876
Encompass_Preferred
Dishwashing liquid uses powerful, grease-cutting ingredients to leave your dishes sparkling clean. Long-lasting suds reduce sink changeover while manually washing pots and pans. High-surfactant formula seeks out greasy soil and cuts through it. The dishwashing liquid surrounds the removed soil and lifts it out of the way to quickly work through larger loads. Multipurpose soap is also powerful enough to clean greasy kitchen surfaces, pretreat stains on clothes and remove grease and oil from metals.
Catalog No. 50-007-90876 Supplier Enterprise Technology Solutions Supplier No. PGC57445
Dishwashing liquid uses powerful, grease-cutting ingredients to leave your dishes sparkling clean. Long-lasting suds reduce sink changeover while manually washing pots and pans. High-surfactant formula seeks out greasy soil and cuts through it. The dishwashing liquid surrounds the removed soil and lifts it out of the way to quickly work through larger loads. Multipurpose soap is also powerful enough to clean greasy kitchen surfaces, pretreat stains on clothes and remove grease and oil from metals.