Learn More
Enterprise Technology Solutions Procter & Gamble Commercial Drain Cleaner,Bio XL,f/Buildup&Back-Up,Microorganisms,Natural,5 Gallons,White
SDP
Catalog No. 5000790213
Encompass_Preferred
DCT Bio Drain XL is designed for optimal maintenance of aqueous environments, such as: drain lines, septic systems, grease traps/interceptors and holding tanks. Specialty formula reduces buildup, prevents back-up and keeps systems free flowing. Cleaner contains microorganisms, selected for exceptional ability to break down organic compounds like oil, grease, proteins, carbohydrates, cellulose and more.
Catalog No. 50-007-90213 Supplier Enterprise Technology Solutions Supplier No. PGC00086
DCT Bio Drain XL is designed for optimal maintenance of aqueous environments, such as: drain lines, septic systems, grease traps/interceptors and holding tanks. Specialty formula reduces buildup, prevents back-up and keeps systems free flowing. Cleaner contains microorganisms, selected for exceptional ability to break down organic compounds like oil, grease, proteins, carbohydrates, cellulose and more.