Learn More
Enterprise Technology Solutions Procter & Gamble Commercial Floor Cleaner,f/Finished Floors,f/Auto Floor Machines,1G,GN
SDP
Catalog No. 5000790247
Encompass_Preferred
Floor cleaner dissolves and removes tough, ground-in dirt from finished floors without leaving a dulling residue. Great for use in restaurants (front of house), salons, medical/dental offices, schools, daycares, offices and churches. Low-sudsing formula makes it an excellent choice for use in automatic floor-scrubbing machines. Cleaner contains no phosphates, nonylphenol ethoxylates, butyl cellosolve, dibutyl phthalate or heavy metals. Recommended dilution ratio is 1:128 for manual and 1:512 for automatic machines.
Catalog No. 50-007-90247 Supplier Enterprise Technology Solutions Supplier No. PGC02037
Floor cleaner dissolves and removes tough, ground-in dirt from finished floors without leaving a dulling residue. Great for use in restaurants (front of house), salons, medical/dental offices, schools, daycares, offices and churches. Low-sudsing formula makes it an excellent choice for use in automatic floor-scrubbing machines. Cleaner contains no phosphates, nonylphenol ethoxylates, butyl cellosolve, dibutyl phthalate or heavy metals. Recommended dilution ratio is 1:128 for manual and 1:512 for automatic machines.