Learn More
Enterprise Technology Solutions Procter & Gamble Commercial Floor Cleaner,f/Finished Floors,f/Auto Machines,1G,3/CT,GN
SDP
Catalog No. 5000790537
Encompass_Preferred
Mr. Clean Professional All-Purpose Floor Cleaner provides excellent cleaning and shine preservation for many types of finished floors, including ceramic and vinyl tile, terrazzo and varnished hardwood. All-purpose cleaner also effectively cleans other surfaces like walls, tables, countertops, restrooms and more. Specially engineered formula creates rapid-collapse suds, making it an ideal choice for automatic floor scrubbing machines.
Catalog No. 50-007-90537 Supplier Enterprise Technology Solutions Supplier No. PGC39949
Mr. Clean Professional All-Purpose Floor Cleaner provides excellent cleaning and shine preservation for many types of finished floors, including ceramic and vinyl tile, terrazzo and varnished hardwood. All-purpose cleaner also effectively cleans other surfaces like walls, tables, countertops, restrooms and more. Specially engineered formula creates rapid-collapse suds, making it an ideal choice for automatic floor scrubbing machines.