Learn More
Enterprise Technology Solutions Procter & Gamble Commercial Floor Cleaner, Spic-n-Span, 1 Gallon, 3/CT, GN
SDP
Catalog No. 5000790678
Encompass_Preferred
Spic and Span Floor Cleaner is a versatile, high-performance, liquid detergent for easy, fast cleaning of foodservice grease found on floors and other areas. This product is specially formulated for foodservice. It cuts tough grease on quarry tile and works on walls, tables, food-contact surfaces and restrooms. Dual-surfactant system emulsifies greasy soils and suspends them so they can be removed. High builder level helps remove grimy tracked-in dirt and delivers excellent cleaning in hard water. Noncorrosive, mildly alkaline formula balances tough cleaning power with employee safety. Concentrated liquid uses 3 fluid oz. per 4-gallon bucket for quarry tile floors. Floor cleaner can be used with chlorine bleach. It is classified by Underwriters Laboratories as to slip-resistance only 43R9. UL.
Catalog No. 50-007-90678 Supplier Enterprise Technology Solutions Supplier No. PGC31569
Spic and Span Floor Cleaner is a versatile, high-performance, liquid detergent for easy, fast cleaning of foodservice grease found on floors and other areas. This product is specially formulated for foodservice. It cuts tough grease on quarry tile and works on walls, tables, food-contact surfaces and restrooms. Dual-surfactant system emulsifies greasy soils and suspends them so they can be removed. High builder level helps remove grimy tracked-in dirt and delivers excellent cleaning in hard water. Noncorrosive, mildly alkaline formula balances tough cleaning power with employee safety. Concentrated liquid uses 3 fluid oz. per 4-gallon bucket for quarry tile floors. Floor cleaner can be used with chlorine bleach. It is classified by Underwriters Laboratories as to slip-resistance only 43R9. UL.