Learn More
Enterprise Technology Solutions Procter & Gamble Commercial Rinse Aid,f/Warewash,All-Temp,Concentrate,1 Gal,2/CT,BE
SDP
Catalog No. 5000830009
Encompass_Preferred
All-Temp Rinse Aid is specially formulated to work in all-temperature warewashing machines. Special conditioning agents reduce surface tension, providing rapid draining and rinsing. Formula counteracts spotting caused by water impurities and dissolved solids. Rinse aid works with additional quaternary ammonium, chlorine-based and iodine-based sanitizer concentrates. Formula delivers exceptional shine every time without the extra work of rewashing, bleaching or polishing. Rinse aid does not contain phthalates, triclosan, benzene, alkylphenols or heavy metals. Keep your machine clean and efficient while doubling the time between needed deliming. Formula contains limescale fighting agents that help prevent calcium buildup and leaves glasses, flatware and other wares shiny and virtually spot-free. Concentrated liquid helps ease dispensing and enables dosing accuracy on a wide range of soils. Rinse aid works in any type of water condition with any dish machine.
Catalog No. 50-008-30009 Supplier Enterprise Technology Solutions Supplier No. PGC71187
All-Temp Rinse Aid is specially formulated to work in all-temperature warewashing machines. Special conditioning agents reduce surface tension, providing rapid draining and rinsing. Formula counteracts spotting caused by water impurities and dissolved solids. Rinse aid works with additional quaternary ammonium, chlorine-based and iodine-based sanitizer concentrates. Formula delivers exceptional shine every time without the extra work of rewashing, bleaching or polishing. Rinse aid does not contain phthalates, triclosan, benzene, alkylphenols or heavy metals. Keep your machine clean and efficient while doubling the time between needed deliming. Formula contains limescale fighting agents that help prevent calcium buildup and leaves glasses, flatware and other wares shiny and virtually spot-free. Concentrated liquid helps ease dispensing and enables dosing accuracy on a wide range of soils. Rinse aid works in any type of water condition with any dish machine.