Learn More
Enterprise Technology Solutions Procter & Gamble Commercial Swiffer Wet Cloths, Sweeper Refills, Disposable, 12 Wipes
SDP
Catalog No. 5000829795
Encompass_Preferred
Swiffer Wet Cloths for the Swiffer Sweeper remove dust instead of just moving it around for dust-free clean. Wet cloths are safe for wood, ceramic, vinyl tile, electronics and other hard surfaces. Do not use wet cloths on unfinished, oiled, or waxed wooden boards, nonsealed tiles or carpeted floors. Wet cloths leave no residue.
Catalog No. 50-008-29795 Supplier Enterprise Technology Solutions Supplier No. PGC08622
Swiffer Wet Cloths for the Swiffer Sweeper remove dust instead of just moving it around for dust-free clean. Wet cloths are safe for wood, ceramic, vinyl tile, electronics and other hard surfaces. Do not use wet cloths on unfinished, oiled, or waxed wooden boards, nonsealed tiles or carpeted floors. Wet cloths leave no residue.