Learn More
Enterprise Technology Solutions Procter & Gamble Commercial Wet-Mop Cloths,Sweeper,Lavender,8"x10",1 Box,White
SDP
Catalog No. 5000829693
Encompass_Preferred
Sweeper Wet Mop Textured Cloths trap and lock dirt deep in cloth for easier, more effective cleaning. Cleaning cloths leave behind scents of lavender and provide lasting freshness for hours. Cleaner formula is safe for all finished floors. Avoid use on unfinished, oiled or waxed wooden boards, non-sealed tiles or carpeted floors.
Catalog No. 50-008-29693 Supplier Enterprise Technology Solutions Supplier No. PGC55311
Sweeper Wet Mop Textured Cloths trap and lock dirt deep in cloth for easier, more effective cleaning. Cleaning cloths leave behind scents of lavender and provide lasting freshness for hours. Cleaner formula is safe for all finished floors. Avoid use on unfinished, oiled or waxed wooden boards, non-sealed tiles or carpeted floors.