Learn More
Med Vet International Proctor Float, 12in, J0058P, Each
Catalog No. 5000584253
Encompass_Preferred
Proctor Float makes it easy to float the first two premolars. Tungsten carbide chips. 12"L.
Catalog No. 50-005-84253 Supplier Med Vet International Supplier No. JOR58P
Proctor Float makes it easy to float the first two premolars. Tungsten carbide chips. 12"L.