Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ ProCulture Warming Plate, Ambient to 37°C, Aluminum

Catalog No. 14513615 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Voltage:
120 V
220 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
14-513-615 220 V
14-513-613 120 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 14-513-615 Supplier Bel-Art™ Supplier No. F370151000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Warming plate is rectangular in shape and is highly accuracte outside the incubator.

  • Increase cell viability during cell passaging with this highly accurate warming plate that keeps cells near an optimal 37°C (±1°C) outside of the incubator.
  • Avoids numerous entries into the CO2 incubator which alter its atmosphere, harming ongoing experiments in the incubated cultures
  • Reduces cell stress and allows enzymes like trypsin to work efficiently
  • Can be used to maintain the warmth of reagents inside the hood
  • Fits easily in cell culture hoods or on the workbench
  • Temperature remains uniform across the entire anodized aluminum warming plate; measures 17.8 D x 20.3 cm W (7 x 8 in.)
  • 15-minute count down timer keeps track of time sensitive applications
  • UV resistant polypropylene housing will not break down under germicidal UV lights
  • Water resistant; safe for cleaning with Ethanol or IPA
  • 220V is CE marked

Compliance

UL approval pending.

Specifications

Top Plate Material Aluminum
Size 7 x 8 in.
Temperature (Metric) Ambient to 37°C
Depth (English) 10 in.
Depth (Metric) 25.4 cm
Height (English) 2.25 in.
Width (English) 9.75 in.
Height (Metric) 5.7 cm
Width (Metric) 24.9 cm
Wattage 22 w
Plug Type EU/UK/AUS/CN
Shape Rectangle
Voltage 220 V
Type Warming Plate
Certifications/Compliance CE
Dimensions (L x W x H) 7 in. x 8 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.