Learn More
Medchemexpress LLC HY-N2343 1mg Medchemexpress, Procyanidin A2 CAS:41743-41-3 Purity:>98%
SDP
Catalog No. 501970385
Encompass_Preferred
Medchemexpress, HY-N2343 1mg Procyanidin A2 CAS:41743-41-3 Procyanidin A2 is a flavonoid found in cranberries and lingonberries, with anti-cancer, antioxidant, antimicrobial and anti-inflammation activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0385 Supplier Medchemexpress LLC Supplier No. HYN23431MG
Add to Cart
Add to Cart
Medchemexpress, HY-N2343 1mg Procyanidin A2 CAS:41743-41-3 Procyanidin A2 is a flavonoid found in cranberries and lingonberries, with anti-cancer, antioxidant, antimicrobial and anti-inflammation activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.